This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03222766_g1
See other TDGF1 GE Assays ›
Gene Symbol:
TDGF1
Gene Name:
teratocarcinoma-derived growth factor 1
Gene Aliases:
-
Chromosome Location:
Chr.22: 52867447 - 52872205 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03222766_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TDGF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080358.1 NP_001073827.1 2 - 3 290 68
GenBank mRNA BT021740.1 - 2 - 3 290 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0