This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03220802_g1
See other RPL18A GE Assays ›
Gene Symbol:
RPL18A
Gene Name:
ribosomal protein L18a
Gene Aliases:
-
Chromosome Location:
Chr.7: 5279456 - 5282849 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03220802_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL18A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033619.2 NP_001028791.1 1 - 2 60 58
GenBank mRNA BC102624.1 - 1 - 2 61 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC102624.1, NM_001033619.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translation
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0