This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03217355_m1
See other CYM GE Assays ›
Gene Symbol:
CYM
Gene Name:
chymosin
Gene Aliases:
CPC, Chy
Chromosome Location:
Chr.3: 32944099 - 32954911 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03217355_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_180994.2 NP_851337.1 5 - 6 671 145
GenBank mRNA AF421161.1 - 5 - 6 632 145
AF421164.1 - 3 - 4 395 145
AF421165.1 - 3 - 4 395 145
EF541122.1 - 5 - 6 646 145
FJ768675.1 - 5 - 6 649 145
J00002.1 - 5 - 6 666 145
J00003.1 - 5 - 6 671 145

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
aspartic-type endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0