This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03216194_m1
See other CERT1 GE Assays ›
Gene Symbol:
CERT1
Gene Name:
ceramide transporter 1
Gene Aliases:
CERT, COL4A3BP
Chromosome Location:
Chr.10: 6793847 - 6925358 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03216194_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174765.3 NP_777190.1 5 - 6 1017 72
GenBank mRNA AF232931.1 - 5 - 6 1018 72
AF232933.1 - 5 - 6 1018 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
in utero embryonic development
heart morphogenesis
ceramide metabolic process
muscle contraction
endoplasmic reticulum organization
signal transduction
cell proliferation
phosphorylation
response to endoplasmic reticulum stress
lipid homeostasis
mitochondrion morphogenesis
intermembrane sphingolipid transfer
ceramide 1-phosphate transport
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0