This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03216190_g1
See other CERT1 GE Assays ›
Gene Symbol:
CERT1
Gene Name:
ceramide transporter 1
Gene Aliases:
CERT, COL4A3BP
Chromosome Location:
Chr.10: 6793847 - 6925358 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03216190_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174765.3 NP_777190.1 1 188 130
GenBank mRNA AF232931.1 - 1 189 130
AF232933.1 - 1 189 130

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_174765.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
in utero embryonic development
heart morphogenesis
ceramide metabolic process
muscle contraction
endoplasmic reticulum organization
signal transduction
cell proliferation
phosphorylation
response to endoplasmic reticulum stress
lipid homeostasis
mitochondrion morphogenesis
intermembrane sphingolipid transfer
ceramide 1-phosphate transport
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0