Assay Details
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174701.2 | NP_777126.1 | 8 - 9 | 716 | 105 | |
GenBank mRNA | AF202997.1 | - | 8 - 9 | 716 | 105 | |
AF238859.1 | - | 8 - 9 | 716 | 105 | ||
BC133473.1 | - | 8 - 9 | 722 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
