This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03215636_m1
See other PRKAR2B GE Assays ›
Gene Symbol:
PRKAR2B
Gene Name:
protein kinase cAMP-dependent type II regulatory subunit beta
Gene Aliases:
-
Chromosome Location:
Chr.4: 48154294 - 48255027 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03215636_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKAR2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174649.2 NP_777074.1 1 - 2 303 58
GenBank mRNA BC151299.1 - 1 - 2 546 58
J05692.1 - 1 - 2 303 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cAMP-dependent protein kinase activity
protein binding
cAMP-dependent protein kinase regulator activity
cAMP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0