This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03213756_gH
See other GZMB GE Assays ›
Gene Symbol:
GZMB
Gene Name:
granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1)
Gene Aliases:
BDMD1, CTLA1
Chromosome Location:
Chr.21: 34741243 - 34743324 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03213756_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GZMB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174296.2 NP_776721.1 2 - 3 224 97
GenBank mRNA AF198965.1 - 2 - 3 224 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0