This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03213713_m1
See other HAPLN1 GE Assays ›
Gene Symbol:
HAPLN1
Gene Name:
hyaluronan and proteoglycan link protein 1
Gene Aliases:
CRTL1, LP
Chromosome Location:
Chr.7: 83535917 - 83616647 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03213713_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAPLN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174288.1 NP_776713.1 1 - 2 79 96
GenBank mRNA U02292.1 - 1 - 2 79 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_174288.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
hyaluronic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0