This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03213303_m1
See other YARS1 GE Assays ›
Gene Symbol:
YARS1
Gene Name:
tyrosyl-tRNA synthetase 1
Gene Aliases:
YARS, tyrS
Chromosome Location:
Chr.2: 120849176 - 120881197 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03213303_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174220.2 NP_776645.1 11 - 12 1428 110
GenBank mRNA AF087021.1 - 11 - 12 1428 110
BC122623.1 - 11 - 12 1426 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

tyrosyl-tRNA aminoacylation
tRNA binding
tyrosine-tRNA ligase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0