This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03212845_m1
See other NPPC GE Assays ›
Gene Symbol:
NPPC
Gene Name:
natriuretic peptide C
Gene Aliases:
-
Chromosome Location:
Chr.2: 119585622 - 119590237 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03212845_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPPC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174125.2 NP_776550.1 2 - 3 540 84
GenBank mRNA BC123399.1 - 2 - 3 448 84
Z48477.1 - 540 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

ossification
growth plate cartilage chondrocyte differentiation
growth plate cartilage chondrocyte proliferation
cGMP biosynthetic process
receptor guanylyl cyclase signaling pathway
receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0