Assay Details
Gene Symbol: DLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174037.2 | NP_776462.1 | 5 - 6 | 842 | 120 | |
GenBank mRNA | AB009278.1 | - | 5 - 6 | 780 | 120 | |
AF181462.1 | - | 5 - 6 | 842 | 120 | ||
BC120429.1 | - | 5 - 6 | 842 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
