This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03211905_m1
See other IL6 GE Assays ›
Gene Symbol:
IL6
Gene Name:
interleukin 6
Gene Aliases:
-
Chromosome Location:
Chr.4: 31454776 - 31459143 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03211905_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173923.2 NP_776348.1 4 - 5 512 115
GenBank mRNA BC123577.1 - 4 - 5 532 115
EU276071.1 - 4 - 5 475 115
X57317.1 - 512 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neutrophil apoptotic process
hepatic immune response
positive regulation of acute inflammatory response
acute-phase response
vascular endothelial growth factor production
positive regulation of vascular endothelial growth factor production
positive regulation of epithelial to mesenchymal transition
response to activity
neuron projection development
positive regulation of chemokine production
positive regulation of interleukin-6 production
negative regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of STAT protein
glucose homeostasis
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of osteoblast differentiation
positive regulation of translation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
regulation of insulin secretion
positive regulation of B cell activation
positive regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
response to glucocorticoid
defense response to virus
positive regulation of glial cell proliferation
regulation of glucagon secretion
interleukin-6-mediated signaling pathway
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
T-helper 17 cell lineage commitment
hepatocyte proliferation
positive regulation of extracellular matrix disassembly
liver regeneration
positive regulation of apoptotic DNA fragmentation
positive regulation of production of miRNAs involved in gene silencing by miRNA
positive regulation of STAT cascade
positive regulation of leukocyte adhesion to vascular endothelial cell
cytokine activity
interleukin-6 receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0