This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba04646133_s1
Gene Symbol:
Bacteria
Gene Name:
OXA-1 family class D beta-lactamase (blaOXA)
Gene Aliases:
Bacteria, blaOXA-1, blaOXA-1042, blaOXA-224, blaOXA-31, blaOXA-320, blaOXA-392, blaOXA-4, blaOXA-47, blaOXA-534, blaOXA-675, blaOXA-796, OXA-1 family class D beta-lactamase OXA, blaOXA
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID Ba04646133_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Set Membership:

Recommended Assay

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0