This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba00005023_po
Gene Symbol:
Bacteria
Gene Name:
D-alanine--(R)-lactate ligase VanA|VanB
Gene Aliases:
Bacteria, D-alanine--(R)-lactate ligase (vanA), D-alanine--(R)-lactate ligase (vanB), vanA,vanB
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID Ba00005023_po
Best Coverage
Size
Availability Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Updated Mapping Information

This pooled assay is composed of the following assays: Ba04646147_s1, Ba04646150_s1

Set Membership:

Recommended Assay Microbe Detection

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0