This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005004_po
Gene Symbol:
Mycoplasma genitalium
Gene Name:
Mycoplasma hominis; Mycoplasma genitalium
Gene Aliases:
Mycoplasma genitalium, Mycoplasma hominis
Chromosome Location:
-
Species:
Mycoplasma genitalium
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005004_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04646251_s1, Ba04646255_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header