This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005001_po
Gene Symbol:
Enterobacter hormaechei
Gene Name:
Klebsiella aerogenes; Enterobacter cloacae complex
Gene Aliases:
Enterobacter hormaechei, Enterobacter kobei, Enterobacter ludwigii, Klebsiella aerogenes, Enterobacter cloacae, Enterobacter asburiae, Lelliottia nimipressuralis
Chromosome Location:
-
Species:
Enterobacter hormaechei
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005001_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04932080_s1, Ba04932087_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0