This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At03402015_g1
See other DCI1 GE Assays ›
Gene Symbol:
DCI1
Gene Name:
delta(3,5),delta(2,4)-dienoyl-CoA isomerase 1
Gene Aliases:
''delta(3, 4)-DIENOYL-, 4)-DIENOYL-COA ISOMERASE 1, 4)-dienoyl-CoA isomerase 1, 4)-dienoyl-CoA isomerase 1'', 5), AT5G43280, ATDCI1, DELTA(2, DELTA(3, MNL12.10, MNL12_10, delta(2, delta(3
Chromosome Location:
Chr.5: 17367827 - 17369510 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At03402015_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085229.1 NP_001078698.1 1 - 2 704 132
GenBank mRNA AK316991.1 - 1 - 2 672 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
fatty acid catabolic process
delta3,5-delta2,4-dienoyl-CoA isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0