This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02545835_s1
See other TIM17-3 GE Assays ›
Gene Symbol:
TIM17-3
Gene Name:
translocase inner membrane subunit 17-3
Gene Aliases:
ARABIDOPSIS THALIANA TRANSLOCASE INNER MEMBRANE SUBUNIT 17-3, ARABIDOPSIS THALIANA TRANSLOCASE INNER MEMBRANE SUBUNIT 17-3, AT5G11690, ATTIM17-3, T22P22.80, T22P22_80, translocase inner membrane subunit 17-3
Chromosome Location:
Chr.5: 3761188 - 3762346 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02545835_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIM17-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121207.4 NP_196730.1 2 809 145

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein import into mitochondrial matrix
protein transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header