This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02543884_s1
See other PFL GE Assays ›
Gene Symbol:
PFL RPS18C
Gene Name:
Ribosomal protein S13/S18 family
S18 ribosomal protein
Gene Aliases:
40S RIBOSOMAL PROTEIN S18, AT1G22780, PFL1, POINTED FIRST LEAVES, POINTED FIRST LEAVES 1, RPS18A, T22J18.5, T22J18_5
AT4G09800, F17A8.150, S18 RIBOSOMAL PROTEIN, S18 ribosomal protein
Chromosome Location:
Chr.1: 8067864 - 8069356 on Build TAIR10.1
Chromosome Location:
Chr.4: 6173712 - 6175200 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02543884_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PFL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_102125.4 NP_173692.1 4 678 134
Gene Symbol: RPS18C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA Z28701.1 - 623 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0