This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02401084_g1
See other SAP130A GE Assays ›
Gene Symbol:
SAP130a SAP130b
Gene Name:
Cleavage and polyadenylation specificity factor (CPSF) A subunit protein
Cleavage and polyadenylation specificity factor (CPSF) A subunit protein
Gene Aliases:
AT3G55200, Arabidopsis thaliana spliceosome-associated protein 130 a, AtSAP130a, spliceosome-associated protein 130 a
AT3G55220, Arabidopsis thaliana spliceosome-associated protein 130 b, AtSAP130b, T26I12.100, spliceosome-associated protein 130 b
Chromosome Location:
Chr.3: 20460329 - 20465201 on Build TAIR10.1
Chromosome Location:
Chr.3: 20466676 - 20471171 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02401084_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SAP130a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339727.1 NP_001319757.1 3 3786 144
NM_001339728.1 NP_001327611.1 3 3603 144
NM_115378.4 NP_567015.1 3 3807 144
Gene Symbol: SAP130b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_115380.3 NP_567016.1 3 3830 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

pollen development
pollen germination
plant ovule development
microgametogenesis
nucleic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0