This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02337644_g1
See other GSTT2 GE Assays ›
Gene Symbol:
GSTT2
Gene Name:
glutathione S-transferase THETA 2
Gene Aliases:
AT5G41240, ATGSTT2, GST10B, K1O13.3, K1O13_3, glutathione S-transferase THETA 2
Chromosome Location:
Chr.5: 16497959 - 16500865 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02337644_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123489.4 NP_198940.3 7 - 8 778 117
GenBank mRNA AY139996.1 - 7 - 8 771 117
BT008361.1 - 7 - 8 724 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

toxin catabolic process
glutathione transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0