This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02336564_gH
See other DMT2 GE Assays ›
Gene Symbol:
DMT2
Gene Name:
DNA methyltransferase 2
Gene Aliases:
AT4G14140, DL3110W, DMT02, DNA METHYLTRANSFERASE 02, DNA METHYLTRANSFERASE 2, DNA methyltransferase 2, FCAALL.111, MET02, MET2
Chromosome Location:
Chr.4: 8145998 - 8152131 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02336564_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DMT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203796.1 NP_001190725.1 1 - 2 1403 172
NM_001340902.1 NP_001319931.1 2 - 3 1537 172
NM_117491.1 NP_193150.1 1 - 2 1403 172

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA mediated transformation
C-5 methylation of cytosine
chromatin binding
DNA (cytosine-5-)-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0