This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02336511_g1
See other AT4G11320 GE Assays ›
Gene Symbol:
AT4G11320
Gene Name:
Papain family cysteine protease
Gene Aliases:
AT4G11320, AtCP2, CP2, F8L21.110, F8L21_110, cysteine protease 2
Chromosome Location:
Chr.4: 6887166 - 6889879 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02336511_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G11320
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340705.1 NP_001328659.1 2 - 3 841 91
NM_117203.4 NP_567377.1 2 - 3 902 91
GenBank mRNA AY035055.1 - 2 - 3 814 91
AY051062.1 - 2 - 3 732 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
proteolysis involved in cellular protein catabolic process
cysteine-type endopeptidase activity
peptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0