This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02335468_g1
See other PAD3 GE Assays ›
Gene Symbol:
PAD3
Gene Name:
Cytochrome P450 superfamily protein
Gene Aliases:
AT3G26830, CYP71B15, PHYTOALEXIN DEFICIENT 3
Chromosome Location:
Chr.3: 9887917 - 9889791 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02335468_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113595.4 NP_189318.1 1 - 2 957 139
GenBank mRNA AK117967.1 - 1 - 2 926 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

defense response
response to water deprivation
response to bacterium
response to insect
indole phytoalexin biosynthetic process
response to abscisic acid
regulation of systemic acquired resistance
camalexin biosynthetic process
defense response to fungus
monooxygenase activity
iron ion binding
dihydrocamalexic acid decarboxylase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
heme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0