This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02335170_g1
See other PGK1 GE Assays ›
Gene Symbol:
PGK1
Gene Name:
phosphoglycerate kinase 1
Gene Aliases:
AT3G12780, phosphoglycerate kinase 1
Chromosome Location:
Chr.3: 4060920 - 4063306 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02335170_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_112114.4 NP_187884.1 2 - 3 666 91
GenBank mRNA AF247558.1 - 2 - 3 527 91
AF360234.1 - 2 - 3 544 91
AF428418.1 - 2 - 3 536 91
AK227225.1 - 2 - 3 545 91
AK316922.1 - 2 - 3 544 91
AY045623.1 - 2 - 3 559 91
AY059841.1 - 2 - 3 543 91
AY062953.1 - 2 - 3 500 91
AY093998.1 - 2 - 3 500 91
AY114638.1 - 2 - 3 500 91
AY126991.1 - 2 - 3 536 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Endogenous Controls Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

carbohydrate kinase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
response to cold
thylakoid membrane organization
reductive pentose-phosphate cycle
galactolipid biosynthetic process
response to cadmium ion
positive regulation of oxidative phosphorylation
mRNA binding
phosphoglycerate kinase activity
protein kinase activity
ATP binding
ADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0