This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02335107_g1
See other MEE67 GE Assays ›
Gene Symbol:
MEE67 AT1G18320
Gene Name:
Mitochondrial import inner membrane translocase subunit Tim17/Tim22/Tim23 family protein
Mitochondrial import inner membrane translocase subunit Tim17/Tim22/Tim23 family protein
Gene Aliases:
AT3G10110, maternal effect embryo arrest 67
AT1G18320, F15H18.28, F15H18_28
Chromosome Location:
Chr.3: 3116110 - 3117894 on Build TAIR10.1
Chromosome Location:
Chr.1: 6304360 - 6305689 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02335107_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEE67
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337857.1 NP_001327144.1 2 - 3 477 149
NM_111846.3 NP_566368.1 2 - 3 477 149
GenBank mRNA BT030038.1 - 2 - 3 362 149
Gene Symbol: AT1G18320
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_101690.3 NP_173268.3 2 - 3 381 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

embryo development ending in seed dormancy
protein import into mitochondrial inner membrane
protein transmembrane transporter activity
mitochondrion targeting sequence binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0