This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02333864_g1
See other UBC10 GE Assays ›
Gene Symbol:
UBC10
Gene Name:
ubiquitin-conjugating enzyme 10
Gene Aliases:
AT5G53300, ubiquitin-conjugating enzyme 10
Chromosome Location:
Chr.5: 21632021 - 21634314 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02333864_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBC10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161304.2 NP_001154776.1 3 - 4 427 140
NM_001203599.1 NP_001190528.1 3 - 4 360 140
NM_124709.2 NP_568788.1 3 - 4 303 140
NM_180850.3 NP_851181.1 3 - 4 421 140
GenBank mRNA AF324718.2 - 3 - 4 291 140
AF325005.2 - 3 - 4 290 140
AF326872.1 - 3 - 4 295 140
AY039566.1 - 3 - 4 299 140
AY065059.1 - 3 - 4 295 140
AY113937.1 - 2 - 3 201 140
AY129488.1 - 2 - 3 201 140
DQ027024.1 - 2 - 3 201 140
L00640.1 - 3 - 4 333 140
Z14991.1 - 333 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
ubiquitin protein ligase binding
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0