This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02326893_g1
See other AT2G43420 GE Assays ›
Gene Symbol:
AT2G43420
Gene Name:
3-beta hydroxysteroid dehydrogenase/isomerase family protein
Gene Aliases:
AT2G43420, T1O24.16
Chromosome Location:
Chr.2: 18031268 - 18035182 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02326893_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT2G43420
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_129903.3 NP_565998.1 5 - 6 998 74
GenBank mRNA AK117930.1 - 5 - 6 903 74
AY045841.1 - 5 - 6 869 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Process(es)

C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-beta-hydroxy-delta5-steroid dehydrogenase activity
protein binding
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
sterol-4-alpha-carboxylate 3-dehydrogenase (decarboxylating) activity
4alpha-carboxy-4beta-methyl-5alpha-cholesta-8-en-3beta-ol:NAD(P)+ 3-oxidoreductase (decarboxylating) activity
4alpha-carboxy-5alpha-cholesta-8-en-3beta-ol:NAD(P)+ 3-dehydrogenase (decarboxylating) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0