This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02324308_g1
See other AT2G38950 GE Assays ›
Gene Symbol:
AT2G38950
Gene Name:
Transcription factor jumonji (jmj) family protein / zinc finger (C5HC2 type) family protein
Gene Aliases:
AT2G38950, T7F6.12, T7F6_12
Chromosome Location:
Chr.2: 16260803 - 16265270 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02324308_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT2G38950
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_129453.4 NP_181429.2 6 - 7 1392 70
GenBank mRNA AY127026.1 - 6 - 7 1284 70
BT001088.1 - 5 - 6 1162 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
regulation of transcription, DNA-templated
histone demethylase activity
histone demethylase activity (H3-trimethyl-K4 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0