This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02322396_g1
See other CSLA07 GE Assays ›
Gene Symbol:
CSLA07
Gene Name:
cellulose synthase like
Gene Aliases:
AT2G35650, ATCSLA07, ATCSLA7, CELLULOSE SYNTHASE LIKE A7, CSLA7, T20F21.16, T20F21_16, cellulose synthase like
Chromosome Location:
Chr.2: 14985470 - 14988362 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02322396_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSLA07
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001336568.1 NP_001325223.1 5 - 6 1132 95
NM_001336569.1 NP_001325221.1 5 - 6 988 95
NM_001336570.1 NP_001325222.1 4 - 5 831 95
NM_129120.4 NP_565813.1 5 - 6 1011 95
GenBank mRNA AJ488284.1 - 5 - 6 993 95
AY059724.1 - 5 - 6 982 95
AY133807.1 - 5 - 6 856 95
KJ138872.1 - 5 - 6 856 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Process(es)

transferase activity, transferring glycosyl groups
glucomannan 4-beta-mannosyltransferase activity
mannan synthase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0