Assay Details
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336567.1 | NP_001325149.1 | 9 - 10 | 1563 | 105 | |
NM_129117.5 | NP_565811.2 | 9 - 10 | 1257 | 105 | ||
GenBank mRNA | AF367246.1 | - | 9 - 10 | 1003 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
