Assay Details
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336567.1 | NP_001325149.1 | 52 - 53 | 6240 | 85 | |
NM_129117.5 | NP_565811.2 | 52 - 53 | 5934 | 85 | ||
GenBank mRNA | AF367246.1 | - | 52 - 53 | 5680 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
