This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02322376_g1
See other MOR1 GE Assays ›
Gene Symbol:
MOR1
Gene Name:
ARM repeat superfamily protein
Gene Aliases:
AT2G35630, GEM1, MICROTUBULE ORGANIZATION 1, T20F21.17, T20F21_17
Chromosome Location:
Chr.2: 14966574 - 14980761 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02322376_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001336567.1 NP_001325149.1 46 - 47 5353 91
NM_129117.5 NP_565811.2 46 - 47 5047 91
GenBank mRNA AF367246.1 - 46 - 47 4793 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
cytokinesis by cell plate formation
mitotic spindle organization
cell plate formation involved in plant-type cell wall biogenesis
establishment or maintenance of microtubule cytoskeleton polarity
microtubule polymerization
microtubule binding
microtubule plus end polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0