Assay Details
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336567.1 | NP_001325149.1 | 2 - 3 | 465 | 61 | |
NM_129117.5 | NP_565811.2 | 2 - 3 | 526 | 61 | ||
GenBank mRNA | AF367246.1 | - | 2 - 3 | 272 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001336567.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
