This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02321122_g1
See other AT5G53330 GE Assays ›
Gene Symbol:
AT5G53330
Gene Name:
Ubiquitin-associated/translation elongation factor EF1B protein
Gene Aliases:
AT5G53330, K19E1.13, K19E1_13
Chromosome Location:
Chr.5: 21639216 - 21640963 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02321122_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G53330
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124712.3 NP_568790.1 2 - 3 278 124
GenBank mRNA AY084524.1 - 2 - 3 277 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
ubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0