This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02320204_g1
See other PGM GE Assays ›
Gene Symbol:
PGM
Gene Name:
phosphoglucomutase
Gene Aliases:
ARABIDOPSIS THALIANA PHOSPHOGLUCOMUTASE, AT5G51820, ATPGMP, MIO24.4, MIO24_4, PGM1, PHOSPHOGLUCOMUTASE, STARCH-FREE 1, STF1, phosphoglucomutase
Chromosome Location:
Chr.5: 21063298 - 21068327 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02320204_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124561.3 NP_199995.1 7 - 8 973 133
GenBank mRNA AF216580.1 - 7 - 8 688 133
AY099708.1 - 7 - 8 644 133
AY128901.1 - 7 - 8 579 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
response to cold
detection of gravity
starch biosynthetic process
magnesium ion binding
phosphoglucomutase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0