This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02317192_g1
See other LBA1 GE Assays ›
Gene Symbol:
LBA1
Gene Name:
RNA helicase
Gene Aliases:
AT5G47010, ATUPF1, LOW-LEVEL BETA-AMYLASE 1, MQD22.15, MQD22_15, UPF1
Chromosome Location:
Chr.5: 19071824 - 19079453 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02317192_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LBA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124072.3 NP_199512.2 14 - 15 2135 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
response to wounding
response to sucrose
salicylic acid mediated signaling pathway
jasmonic acid mediated signaling pathway
sugar mediated signaling pathway
RNA interference
defense response to bacterium
long-day photoperiodism
DNA binding
RNA binding
RNA helicase activity
mRNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0