This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02315417_g1
See other CBP20 GE Assays ›
Gene Symbol:
CBP20
Gene Name:
CAP-binding protein 20
Gene Aliases:
AT5G44200, ATCBP20, CAP-binding protein 20, MLN1.12, MLN1_12
Chromosome Location:
Chr.5: 17801737 - 17804203 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02315417_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CBP20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085236.2 NP_001078705.1 3 - 4 470 123
NM_123787.4 NP_199233.1 2 - 3 468 123
GenBank mRNA AF140219.1 - 2 - 3 289 123
AK227850.1 - 2 - 3 424 123
BT004813.1 - 2 - 3 237 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via endonucleolytic cleavage and ligation
mRNA splicing, via spliceosome
RNA metabolic process
primary miRNA processing
mRNA cis splicing, via spliceosome
RNA cap binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0