This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02314851_g1
See other C-NAD-MDH2 GE Assays ›
Gene Symbol:
c-NAD-MDH2
Gene Name:
Lactate/malate dehydrogenase family protein
Gene Aliases:
AT5G43330, MWF20.2, MWF20_2, cytosolic-NAD-dependent malate dehydrogenase 2
Chromosome Location:
Chr.5: 17390381 - 17392741 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02314851_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: c-NAD-MDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123699.4 NP_199147.1 6 - 7 1074 85
GenBank mRNA AY072137.1 - 6 - 7 976 85
AY091220.1 - 6 - 7 903 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
tricarboxylic acid cycle
oxaloacetate metabolic process
malate metabolic process
NADH metabolic process
protein binding
L-malate dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header