This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02314331_g1
See other ARC6 GE Assays ›
Gene Symbol:
ARC6
Gene Name:
Chaperone DnaJ-domain superfamily protein
Gene Aliases:
ACCUMULATION AND REPLICATION OF CHLOROPLASTS 6, AT5G42480, MDH9.18, MDH9_18
Chromosome Location:
Chr.5: 16985096 - 16988796 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02314331_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARC6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344453.1 NP_001331076.1 3 - 4 1135 140
NM_123613.4 NP_199063.1 3 - 4 1196 140
GenBank mRNA AY091075.1 - 3 - 4 1110 140
AY150490.1 - 3 - 4 997 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chloroplast organization
chloroplast fission
protein binding
protein self-association

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0