This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02313957_g1
See other AT5G41950 GE Assays ›
Gene Symbol:
AT5G41950
Gene Name:
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases:
AT5G41950, MJC20.5, MJC20_5
Chromosome Location:
Chr.5: 16785680 - 16789676 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02313957_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G41950
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123560.4 NP_199010.1 2 - 3 773 89
GenBank mRNA AY085561.1 - 2 - 3 739 89
AY128348.1 - 2 - 3 683 89
BT030345.1 - 2 - 3 628 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
vesicle-mediated transport
root hair elongation
root hair cell tip growth
root hair cell development
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0