This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02313876_g1
See other CIP1 GE Assays ›
Gene Symbol:
CIP1
Gene Name:
COP1-interactive protein 1
Gene Aliases:
AT5G41790, COP1-interactive protein 1, K16L22.7, K16L22_7
Chromosome Location:
Chr.5: 16725753 - 16732847 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02313876_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344421.1 NP_001330520.1 4 - 5 5248 62
NM_123544.3 NP_198994.2 2 - 3 4772 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to osmotic stress
response to light stimulus
response to abscisic acid
positive regulation of abscisic acid-activated signaling pathway
regulation of protein import into nucleus
cellular response to abscisic acid stimulus
actin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header