This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02313357_g1
See other TOM20-4 GE Assays ›
Gene Symbol:
TOM20-4
Gene Name:
translocase of outer membrane 20-4
Gene Aliases:
AT5G40930, MMG1.2, MMG1_2, translocase of outer membrane 20-4
Chromosome Location:
Chr.5: 16404457 - 16406079 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02313357_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOM20-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344374.1 NP_001330863.1 4 - 5 459 79
NM_123458.5 NP_198909.1 4 - 5 459 79
GenBank mRNA AF462861.1 - 4 - 5 456 79
AK117822.1 - 4 - 5 444 79
AY141998.1 - 4 - 5 393 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to mitochondrion
protein import into mitochondrial outer membrane
protein binding
P-P-bond-hydrolysis-driven protein transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0