This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02313354_g1
See other TOM20-4 GE Assays ›
Gene Symbol:
TOM20-4
Gene Name:
translocase of outer membrane 20-4
Gene Aliases:
AT5G40930, MMG1.2, MMG1_2, translocase of outer membrane 20-4
Chromosome Location:
Chr.5: 16404457 - 16406079 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02313354_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOM20-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344374.1 NP_001330863.1 1 - 2 162 70
NM_123458.5 NP_198909.1 1 - 2 162 70
GenBank mRNA AF462861.1 - 1 - 2 159 70
AK117822.1 - 1 - 2 147 70
AY141998.1 - 1 - 2 96 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to mitochondrion
protein import into mitochondrial outer membrane
protein binding
P-P-bond-hydrolysis-driven protein transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0