Assay Details
Gene Symbol: ERA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344319.1 | NP_001331518.1 | 12 - 13 | 1312 | 68 | |
NM_123392.2 | NP_198844.1 | 13 - 14 | 1264 | 68 | ||
GenBank mRNA | AF214106.1 | - | 13 - 14 | 1216 | 68 | |
BT030457.1 | - | 13 - 14 | 1216 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
