This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02312173_g1
See other ABCB27 GE Assays ›
Gene Symbol:
ABCB27
Gene Name:
transporter associated with antigen processing protein 2
Gene Aliases:
ALS1, ALUMINUM SENSITIVE 1, ARABIDOPSIS THALIANA TRANSPORTER ASSOCIATED WITH ANTIGEN PROCESSING PROTEIN 2, AT5G39040, ATP-binding cassette B27, ATTAP2, AtALS1, MXF12.50, MXF12_50, TAP2, transporter associated with antigen processing protein 2
Chromosome Location:
Chr.5: 15625460 - 15629835 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02312173_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCB27
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344283.1 NP_001332027.1 1 - 2 225 89
NM_123266.8 NP_198720.2 1 - 2 289 89
GenBank mRNA AK229810.1 - 1 - 2 171 89
DQ469798.1 - 1 - 2 89 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

response to aluminum ion
transmembrane transport
ATP binding
ATPase activity
transmembrane transporter activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0