Assay Details
| Gene Symbol: AT5G34885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122878.2 | NP_568524.1 | 1 | 438 | 102 | |
| Gene Symbol: AT5G34881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148015.2 | NP_680320.1 | 1 | 102 | ||
More Information
Set Membership: |
|

AT5G34885
Genomic Map