This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02310938_g1
See other FRS10 GE Assays ›
Gene Symbol:
FRS10
Gene Name:
FAR1-related sequence 10
Gene Aliases:
AT5G28530, FAR1-related sequence 10, T26D3.9, T26D3_9
Chromosome Location:
Chr.5: 10524411 - 10527566 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02310938_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FRS10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344073.1 NP_001331636.1 1 - 2 1281 67
NM_001344074.1 NP_001331633.1 1 - 2 1286 67
NM_001344075.1 NP_001331634.1 2 - 3 1099 67
NM_001344076.1 NP_001331635.1 2 - 3 1099 67
NM_122736.2 NP_198205.2 1 - 2 1022 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0