This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02310556_g1
See other RECQSIM GE Assays ›
Gene Symbol:
RECQSIM
Gene Name:
RECQ helicase SIM
Gene Aliases:
AT5G27680, RECQ helicase SIM, T1G16.10, T1G16_10
Chromosome Location:
Chr.5: 9793745 - 9799149 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02310556_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RECQSIM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344027.1 NP_001330792.1 12 - 13 2578 89
NM_001344028.1 NP_001330796.1 13 - 14 2477 89
NM_001344029.1 NP_001330794.1 13 - 14 2472 89
NM_001344030.1 NP_001330793.1 13 - 14 2304 89
NM_001344031.1 NP_001330795.1 11 - 12 2309 89
NM_122650.5 NP_568499.1 12 - 13 2578 89
GenBank mRNA AJ404475.1 - 12 - 13 2119 89
AY059754.1 - 12 - 13 2390 89
AY133823.1 - 12 - 13 2066 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA unwinding involved in DNA replication
DNA repair
DNA recombination
DNA duplex unwinding
nucleic acid binding
DNA helicase activity
four-way junction helicase activity
3'-5' DNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0